Cyclobutyl-3-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086794-1G 086794-2G 086794-5G
102850-666EA 672.31 USD
102850-666 102850-668 102850-670
Cyclobutyl-3-methoxyphenyl ketone ≥97%
Cyclobutyl-3-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD03841249
CAS Number: 898790-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR