Cyclobutyl-2-methoxyphenyl ketone ≥97%
Supplier: Matrix Scientific
086793-1G
086793-2G
086793-5G
102850-660EA
672.31
USD
102850-660
102850-662
102850-664
Cyclobutyl-2-methoxyphenyl ketone ≥97%
Cyclobutyl-2-methoxyphenyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₄O₂ |
MDL Number:
MFCD03841248 CAS Number: 898790-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...