Cyclobutyl-3-methylphenyl ketone ≥97%

Supplier: Matrix Scientific

086791-1G 086791-2G 086791-5G
102850-648EA 672.31 USD
102850-648 102850-650 102850-652
Cyclobutyl-3-methylphenyl ketone ≥97%
Cyclobutyl-3-methylphenyl ketone

Hazard: Irritant

Formula: C₁₂H₁₄O
MDL Number: MFCD03841246
CAS Number: 898790-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR