Cyclobutyl-2-methylphenyl ketone ≥97%
Supplier: Matrix Scientific
086790-1G
086790-2G
086790-5G
102850-642EA
672.31
USD
102850-642
102850-644
102850-646
Cyclobutyl-2-methylphenyl ketone ≥97%
Cyclobutyl-2-methylphenyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₄O |
MDL Number:
MFCD03841245 CAS Number: 898790-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...