2-Chloro-4-fluorophenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086789-1G
086789-2G
086789-5G
102850-636EA
672.31
USD
102850-636
102850-638
102850-640
2-Chloro-4-fluorophenyl cyclopropyl ketone ≥97%
2-Chloro-4-fluorophenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₀H₈ClFO |
MDL Number:
MFCD03841233 CAS Number: 898790-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...