Cyclopropyl-4-trifluoromethylphenyl ketone ≥97%
Supplier: Matrix Scientific
086787-1G
086787-2G
086787-5G
102850-624EA
672.31
USD
102850-624
102850-626
102850-628
Cyclopropyl-4-trifluoromethylphenyl ketone ≥97%
Cyclopropyl-4-trifluoromethylphenyl ketone
Hazard: Irritant
Formula:
C₁₁H₉F₃O |
MDL Number:
MFCD03841231 CAS Number: 62587-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...