Cyclopropyl-3-trifluoromethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086786-1G 086786-2G 086786-5G
102850-618EA 672.31 USD
102850-618 102850-620 102850-622
Cyclopropyl-3-trifluoromethylphenyl ketone ≥97%
Cyclopropyl-3-trifluoromethylphenyl ketone

Hazard: Irritant

Formula: C₁₁H₉F₃O
MDL Number: MFCD03841230
CAS Number: 38675-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR