2-Fluorophenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086785-1G
086785-2G
086785-5G
102850-612EA
1009.23
USD
102850-612
102850-614
102850-616
2-Fluorophenyl cyclopropyl ketone ≥97%
2-Fluorophenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₀H₉FO |
MDL Number:
MFCD03841228 CAS Number: 141030-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...