2-Chlorophenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086784-1G
086784-2G
086784-5G
102850-606EA
672.31
USD
102850-606
102850-608
102850-610
2-Chlorophenyl cyclopropyl ketone ≥97%
2-Chlorophenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₀H₉ClO |
MDL Number:
MFCD03841227 CAS Number: 149914-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...