3-Chloro-4-fluorophenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086783-1G
086783-2G
086783-5G
102850-600EA
672.31
USD
102850-600
102850-602
102850-604
3-Chloro-4-fluorophenyl cyclopropyl ketone ≥97%
3-Chloro-4-fluorophenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₀H₈ClFO |
MDL Number:
MFCD03841226 CAS Number: 898790-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...