4-Bromo-3-fluorophenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086782-1G
086782-2G
086782-5G
102850-594EA
1009.23
USD
102850-594
102850-596
102850-598
4-Bromo-3-fluorophenyl cyclopropyl ketone ≥97%
4-Bromo-3-fluorophenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₀H₈BrFO |
MDL Number:
MFCD03841224 CAS Number: 898790-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...