Cyclopropyl-3,5-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086781-1G 086781-2G 086781-5G
102850-588EA 672.31 USD
102850-588 102850-590 102850-592
Cyclopropyl-3,5-dimethylphenyl ketone ≥97%
Cyclopropyl-3,5-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₂H₁₄O
MDL Number: MFCD03841223
CAS Number: 150668-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR