Cyclopropyl-2,6-dimethylphenyl ketone ≥97%
Supplier: Matrix Scientific
086780-1G
086780-2G
086780-5G
102850-582EA
672.31
USD
102850-582
102850-584
102850-586
Cyclopropyl-2,6-dimethylphenyl ketone ≥97%
Cyclopropyl-2,6-dimethylphenyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₄O |
MDL Number:
MFCD03841222 CAS Number: 870002-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...