4-Carboethoxyphenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086778-1G
086778-2G
086778-5G
102850-570EA
672.31
USD
102850-570
102850-572
102850-574
4-Carboethoxyphenyl cyclopropyl ketone ≥97%
4-Carboethoxyphenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₃H₁₄O₃ |
MDL Number:
MFCD03841215 CAS Number: 863769-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...