3-Carboethoxyphenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086777-1G
086777-2G
086777-5G
102850-564EA
672.31
USD
102850-564
102850-566
102850-568
3-Carboethoxyphenyl cyclopropyl ketone ≥97%
3-Carboethoxyphenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₃H₁₄O₃ |
MDL Number:
MFCD03841214 CAS Number: 878745-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...