2-Carboethoxyphenyl cyclopropyl ketone ≥97%
Supplier: Matrix Scientific
086776-1G
086776-2G
086776-5G
102850-558EA
672.31
USD
102850-558
102850-560
102850-562
2-Carboethoxyphenyl cyclopropyl ketone ≥97%
2-Carboethoxyphenyl cyclopropyl ketone
Hazard: Irritant
Formula:
C₁₃H₁₄O₃ |
MDL Number:
MFCD03841213 CAS Number: 898789-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...