3-Cyanophenyl cyclopropyl ketone ≥97%

Supplier: Matrix Scientific

086775-1G 086775-2G 086775-5G
102850-552EA 672.31 USD
102850-552 102850-554 102850-556
3-Cyanophenyl cyclopropyl ketone ≥97%
3-Cyanophenyl cyclopropyl ketone

Hazard: Irritant

Formula: C₁₁H₉NO
MDL Number: MFCD03841211
CAS Number: 898789-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR