2-Cyanophenyl cyclopropyl ketone ≥97%

Supplier: Matrix Scientific

086774-1G 086774-2G 086774-5G
102850-546EA 672.31 USD
102850-546 102850-548 102850-550
2-Cyanophenyl cyclopropyl ketone ≥97%
2-Cyanophenyl cyclopropyl ketone

Hazard: Irritant

Formula: C₁₁H₉NO
MDL Number: MFCD03841210
CAS Number: 898789-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR