Cyclopropyl-3-methoxyphenyl ketone ≥97%
Supplier: Matrix Scientific
086773-1G
086773-2G
086773-5G
102850-540EA
672.31
USD
102850-540
102850-542
102850-544
Cyclopropyl-3-methoxyphenyl ketone ≥97%
Cyclopropyl-3-methoxyphenyl ketone
Hazard: Irritant
Formula:
C₁₁H₁₂O₂ |
MDL Number:
MFCD03841209 CAS Number: 104271-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...