Cyclopropyl-2-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086772-1G 086772-2G 086772-5G
102850-534EA 1009.23 USD
102850-534 102850-536 102850-538
Cyclopropyl-2-methoxyphenyl ketone ≥97%
Cyclopropyl-2-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD03841208
CAS Number: 400614-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR