Cyclopropyl-3-methylphenyl ketone ≥97%
Supplier: Matrix Scientific
Synonyms:
Cyclopropyl 3-methylphenyl ketone
086771-1G
086771-2G
086771-5G
102850-528EA
672.31
USD
102850-528
102850-530
102850-532
Cyclopropyl-3-methylphenyl ketone ≥97%
Cyclopropyl-3-methylphenyl ketone
Hazard: Irritant
Formula:
C₁₁H₁₂O MW: 160.22 g/mol |
MDL Number:
MFCD03841207 CAS Number: 150668-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...