Cyclopropyl-2-methylphenyl ketone ≥95%

Supplier: Matrix Scientific

086770-1G 086770-2G 086770-5G
102850-522EA 672.31 USD
102850-522 102850-524 102850-526
Cyclopropyl-2-methylphenyl ketone ≥95%
Cyclopropyl-2-methylphenyl ketone

Hazard: Irritant

Formula: C₁₁H₁₂O
MDL Number: MFCD03841206
CAS Number: 39615-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR