2',4'-Difluoro-2,2-dimethylpropiophenone ≥97%
Supplier: Matrix Scientific
086769-1G
086769-2G
086769-5G
102850-516EA
672.31
USD
102850-516
102850-518
102850-520
2',4'-Difluoro-2,2-dimethylpropiophenone ≥97%
2',4'-Difluoro-2,2-dimethylpropiophenone
Hazard: Irritant
Formula:
C₁₁H₁₂F₂O |
MDL Number:
MFCD03841202 CAS Number: 898789-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...