4'-Bromo-2,2-dimethyl-2'-fluoropropiophenone ≥97%

Supplier: Matrix Scientific

086768-1G 086768-2G 086768-5G
102850-510EA 1009.23 USD
102850-510 102850-512 102850-514
4'-Bromo-2,2-dimethyl-2'-fluoropropiophenone ≥97%
4'-Bromo-2,2-dimethyl-2'-fluoropropiophenone

Hazard: Irritant

Formula: C₁₁H₁₂BrFO
MDL Number: MFCD03841193
CAS Number: 898766-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR