4'-Bromo-2,2-dimethylpropiophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Bromophenyl)-2,2-dimethylpropan-1-one
086761-1G
086761-2G
086761-5G
102850-468EA
615.38
USD
102850-468
102850-470
102850-472
4'-Bromo-2,2-dimethylpropiophenone ≥95%
4'-Bromo-2,2-dimethylpropiophenone
Hazard: Irritant
Formula:
C₁₁H₁₃BrO MW: 241.13 g/mol |
MDL Number:
MFCD03841175 CAS Number: 30314-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...