3'-Bromo-2,2-dimethylpropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3-Bromophenyl)-2,2-dimethylpropan-1-one

086760-1G 086760-2G 086760-5G
102850-462EA 966.15 USD
102850-462 102850-464 102850-466
3'-Bromo-2,2-dimethylpropiophenone ≥97%
3'-Bromo-2,2-dimethylpropiophenone

Hazard: Irritant

Formula: C₁₁H₁₃BrO
MW: 241.13 g/mol
MDL Number: MFCD03841174
CAS Number: 2416-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR