2'-Carboethoxy-2,2-dimethylpropiophenone ≥97%

Supplier: Matrix Scientific

086758-1G 086758-2G 086758-5G
102850-450EA 672.31 USD
102850-450 102850-452 102850-454
2'-Carboethoxy-2,2-dimethylpropiophenone ≥97%
2'-Carboethoxy-2,2-dimethylpropiophenone

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD03841170
CAS Number: 898766-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR