3'-Cyano-2,2-dimethylpropiophenone ≥97%

Supplier: Matrix Scientific

086757-1G 086757-2G 086757-5G
102850-444EA 1009.23 USD
102850-444 102850-446 102850-448
3'-Cyano-2,2-dimethylpropiophenone ≥97%
3'-Cyano-2,2-dimethylpropiophenone

Hazard: Irritant

Formula: C₁₂H₁₃NO
MDL Number: MFCD03841168
CAS Number: 898766-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR