2,2-Dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086748-1G
086748-2G
086748-5G
102850-390EA
1009.23
USD
102850-390
102850-392
102850-394
2,2-Dimethylbutyrophenone ≥97%
2,2-Dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₂H₁₆O |
MDL Number:
MFCD00102052 CAS Number: 829-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...