2'-Chloro-3,3-dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086747-1G
086747-2G
086747-5G
102850-384EA
672.31
USD
102850-384
102850-386
102850-388
2'-Chloro-3,3-dimethylbutyrophenone ≥97%
2'-Chloro-3,3-dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₂H₁₅ClO |
MDL Number:
MFCD07699549 CAS Number: 898764-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...