4'-Chloro-3,3-dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086745-1G
086745-2G
086745-5G
102850-372EA
672.31
USD
102850-372
102850-374
102850-376
4'-Chloro-3,3-dimethylbutyrophenone ≥97%
4'-Chloro-3,3-dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₂H₁₅ClO |
MDL Number:
MFCD03841091 CAS Number: 60851-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...