4'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086744-1G
086744-2G
086744-5G
102850-366EA
672.31
USD
102850-366
102850-368
102850-370
4'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%
4'-Carboethoxy-3,3-dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₅H₂₀O₃ |
MDL Number:
MFCD03841085 CAS Number: 684271-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...