3'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086743-1G
086743-2G
086743-5G
102850-360EA
672.31
USD
102850-360
102850-362
102850-364
3'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%
3'-Carboethoxy-3,3-dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₅H₂₀O₃ |
MDL Number:
MFCD03841084 CAS Number: 898764-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...