2'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%

Supplier: Matrix Scientific

086742-1G 086742-2G 086742-5G
102850-354EA 672.31 USD
102850-354 102850-356 102850-358
2'-Carboethoxy-3,3-dimethylbutyrophenone ≥97%
2'-Carboethoxy-3,3-dimethylbutyrophenone

Hazard: Irritant

Formula: C₁₅H₂₀O₃
MDL Number: MFCD03841083
CAS Number: 898764-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR