3,3-Dimethyl-4'-methoxybutyrophenone ≥97%
Supplier: Matrix Scientific
086741-1G
086741-2G
086741-5G
102850-348EA
672.31
USD
102850-348
102850-350
102850-352
3,3-Dimethyl-4'-methoxybutyrophenone ≥97%
3,3-Dimethyl-4'-methoxybutyrophenone
Hazard: Irritant
Formula:
C₁₃H₁₈O₂ |
MDL Number:
MFCD03841079 CAS Number: 85157-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...