3-(4-Cyanophenyl)cyclohexanone ≥97%
Supplier: Matrix Scientific
086736-1G
086736-2G
086736-5G
102850-318EA
886.15
USD
102850-318
102850-320
102850-322
3-(4-Cyanophenyl)cyclohexanone ≥97%
3-(4-Cyanophenyl)cyclohexanone
Hazard: Irritant
Formula:
C₁₃H₁₃NO Melting Pt: 70…72 °C |
MDL Number:
MFCD03841072 CAS Number: 123732-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...