3-(4-Ethylphenyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

086735-1G 086735-2G 086735-5G
102850-312EA 347.69 USD
102850-312 102850-314 102850-316
3-(4-Ethylphenyl)cyclohexanone ≥97%
3-(4-Ethylphenyl)cyclohexanone

Hazard: Irritant

Formula: C₁₄H₁₈O
MW: 202.3 g/mol
MDL Number: MFCD03841068
CAS Number: 898785-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR