4'-Methyl-4-chlorobutyrophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-1-(p-tolyl)butan-1-one
086734-1G
086734-2G
086734-5G
102850-306EA
347.69
USD
102850-306
102850-308
102850-310
4'-Methyl-4-chlorobutyrophenone ≥95%
4'-Methyl-4-chlorobutyrophenone
Hazard: Irritant
Formula:
C₁₁H₁₃ClO MW: 196.673 g/mol |
MDL Number:
MFCD00018999 CAS Number: 38425-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...