3-Chloro-1-(3-methylphenyl)-1-oxopropane ≥97%

Supplier: Matrix Scientific

086733-1G 086733-2G 086733-5G
102850-300EA 661.54 USD
102850-300 102850-302 102850-304
3-Chloro-1-(3-methylphenyl)-1-oxopropane ≥97%
3-Chloro-1-(3-methylphenyl)-1-oxopropane

Hazard: Irritant

Formula: C₁₀H₁₁ClO
MDL Number: MFCD03841051
CAS Number: 898785-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR