4-Chloro-1-(2-methylphenyl)-1-oxobutane ≥95%

Supplier: Matrix Scientific

086732-1G 086732-2G 086732-5G
102850-294EA 592.31 USD
102850-294 102850-296 102850-298
4-Chloro-1-(2-methylphenyl)-1-oxobutane ≥95%
4-Chloro-1-(2-methylphenyl)-1-oxobutane

Hazard: Irritant

Formula: C₁₁H₁₃ClO
MDL Number: MFCD03841047
CAS Number: 40877-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR