3-Chloro-1-(2-methylphenyl)-1-oxopropane ≥97%
Supplier: Matrix Scientific
086731-1G
086731-2G
086731-5G
102850-288EA
592.31
USD
102850-288
102850-290
102850-292
3-Chloro-1-(2-methylphenyl)-1-oxopropane ≥97%
3-Chloro-1-(2-methylphenyl)-1-oxopropane
Hazard: Irritant
Formula:
C₁₀H₁₁ClO |
MDL Number:
MFCD03841046 CAS Number: 898785-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...