3-(2,6-Dimethylphenyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

086730-1G 086730-2G 086730-5G
102850-282EA 592.31 USD
102850-282 102850-284 102850-286
3-(2,6-Dimethylphenyl)cyclohexanone ≥97%
3-(2,6-Dimethylphenyl)cyclohexanone

Hazard: Irritant

Formula: C₁₄H₁₈O
MDL Number: MFCD03841044
CAS Number: 486397-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR