Ethyl-4-(3-oxocyclohexyl)benzoate ≥97%
Supplier: Matrix Scientific
086729-1G
086729-2G
086729-5G
102850-276EA
886.15
USD
102850-276
102850-278
102850-280
Ethyl-4-(3-oxocyclohexyl)benzoate ≥97%
Ethyl-4-(3-oxocyclohexyl)benzoate
Hazard: Irritant
Formula:
C₁₅H₁₈O₃ |
MDL Number:
MFCD03841041 CAS Number: 131379-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...