Ethyl-3-(3-oxocyclohexyl)benzoate ≥97%
Supplier: Matrix Scientific
086728-1G
086728-2G
086728-5G
102850-270EA
886.15
USD
102850-270
102850-272
102850-274
Ethyl-3-(3-oxocyclohexyl)benzoate ≥97%
Ethyl-3-(3-oxocyclohexyl)benzoate
Hazard: Irritant
Formula:
C₁₅H₁₈O₃ |
MDL Number:
MFCD03841040 CAS Number: 284022-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...