4'-Iodo-2-(2-methoxyphenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086726-1G 086726-2G 086726-5G
102850-258EA 592.31 USD
102850-258 102850-260 102850-262
4'-Iodo-2-(2-methoxyphenyl)acetophenone ≥97%
4'-Iodo-2-(2-methoxyphenyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₃IO₂
MDL Number: MFCD02260776
CAS Number: 898784-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR