4'-Bromo-2-(4-fluorophenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086725-1G 086725-5G
102850-252EA 347.69 USD
102850-252 102850-256
4'-Bromo-2-(4-fluorophenyl)acetophenone ≥97%
4'-Bromo-2-(4-fluorophenyl)acetophenone

Hazard: Irritant

Formula: C₁₄H₁₀BrFO
MDL Number: MFCD02260764
CAS Number: 107028-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR