4'-Bromo-2-(2-fluorophenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086724-1G
086724-2G
086724-5G
102850-246EA
886.15
USD
102850-246
102850-248
102850-250
4'-Bromo-2-(2-fluorophenyl)acetophenone ≥97%
4'-Bromo-2-(2-fluorophenyl)acetophenone
Hazard: Irritant
Formula:
C₁₄H₁₀BrFO |
MDL Number:
MFCD02260758 CAS Number: 898784-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...