2-(3-Cyanophenyl)-4'-trifluoromethylacetophenone ≥97%

Supplier: Matrix Scientific

086721-1G 086721-2G 086721-5G
102850-228EA 592.31 USD
102850-228 102850-230 102850-232
2-(3-Cyanophenyl)-4'-trifluoromethylacetophenone ≥97%
2-(3-Cyanophenyl)-4'-trifluoromethylacetophenone

Hazard: Irritant

Formula: C₁₆H₁₀F₃NO
MDL Number: MFCD02260752
CAS Number: 898784-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR