2-(4-Bromophenyl)-2'-trifluoromethylacetophenone ≥97%

Supplier: Matrix Scientific

086719-1G 086719-2G 086719-5G
102850-216EA 592.31 USD
102850-216 102850-218 102850-220
2-(4-Bromophenyl)-2'-trifluoromethylacetophenone ≥97%
2-(4-Bromophenyl)-2'-trifluoromethylacetophenone

Hazard: Irritant

Formula: C₁₅H₁₀BrF₃O
MDL Number: MFCD02260735
CAS Number: 898784-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR