2-(3-Bromophenyl)-3'-trifluoromethylacetophenone ≥97%
Supplier: Matrix Scientific
086718-1G
086718-2G
086718-5G
102850-210EA
592.31
USD
102850-210
102850-212
102850-214
2-(3-Bromophenyl)-3'-trifluoromethylacetophenone ≥97%
2-(3-Bromophenyl)-3'-trifluoromethylacetophenone
Hazard: Irritant
Formula:
C₁₅H₁₀BrF₃O |
MDL Number:
MFCD02260733 CAS Number: 898784-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...